siRNA Details
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172554.2 | NP_766142.2 | 17 | 1851 | ||
GenBank mRNA | AK039575.1 | - | 17 | 1852 | ||
AK046891.1 | - | 17 | 1793 | |||
AK122406.1 | - | 16 | 2796 | |||
BC027101.1 | - | 17 | 1739 | |||
BC117788.1 | - | 17 | 1793 | 40092137 | ||
BC117789.2 | - | 17 | 1793 | 40092140 |
