siRNA Details
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172554.2 | NP_766142.2 | 15 | 1632 | ||
GenBank mRNA | AK039575.1 | - | 15 | 1633 | ||
AK046891.1 | - | 15 | 1574 | |||
AK122406.1 | - | 14 | 2577 | |||
BC027101.1 | - | 15 | 1520 | |||
BC117788.1 | - | 15 | 1574 | 40092137 | ||
BC117789.2 | - | 15 | 1574 | 40092140 |
