siRNA Details
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172554.2 | NP_766142.2 | 12, 13 | 1359 | ||
GenBank mRNA | AK039575.1 | - | 12, 13 | 1360 | ||
AK046891.1 | - | 12, 13 | 1301 | |||
AK122406.1 | - | 11, 12 | 2304 | |||
BC027101.1 | - | 12, 13 | 1247 | |||
BC117788.1 | - | 12, 13 | 1301 | 40092137 | ||
BC117789.2 | - | 12, 13 | 1301 | 40092140 |
