siRNA Details
Gene Symbol: Ccdc171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081012.1 | NP_001074481.1 | 16, 17 | 2382 | ||
GenBank mRNA | AK039282.1 | - | 3, 4 | 500 | ||
BC042767.1 | - | 4, 5 | 625 | |||
BC172165.1 | - | 15, 16 | 2173 |
