siRNA Details
Gene Symbol: Ttll5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081423.2 | NP_001074892.2 | 27 | 2907 | ||
GenBank mRNA | AB093278.1 | - | 27 | 2840 | ||
AK009255.1 | - | 11 | 1285 | |||
AK031470.1 | - | 8 | 979 | |||
AM690748.1 | - | 26 | 2672 | |||
BC030360.1 | - | 2 | 70 | |||
BC035276.1 | - | 26 | 2724 |
