siRNA Details
Gene Symbol: Ttll5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081423.2 | NP_001074892.2 | 13, 14 | 1366 | ||
GenBank mRNA | AB093278.1 | - | 13, 14 | 1299 | ||
AK029125.1 | - | 12, 13 | 3011 | |||
AM690748.1 | - | 12, 13 | 1131 | |||
BC035276.1 | - | 12, 13 | 1241 |
