siRNA Details
Gene Symbol: Iqcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177128.4 | NP_796102.2 | 5 | 501 | ||
GenBank mRNA | AK078554.1 | - | 5 | 494 | ||
AK165220.1 | - | 5 | 485 | |||
BC029084.1 | - | 5 | 441 | 3157011 |
