siRNA Details
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039376.2 | NP_001034465.2 | 2, 3 | 265 | ||
NM_001289701.1 | NP_001276630.1 | 2, 3 | 265 | |||
NM_001289702.1 | NP_001276631.1 | 2, 3 | 265 | |||
GenBank mRNA | AK051209.1 | - | 2, 3 | 343 | ||
AK083867.1 | - | 2, 3 | 340 | |||
AK141917.1 | - | 2, 3 | 391 | |||
BC028274.1 | - | 2, 3 | 275 | |||
BC141185.1 | - | 2, 3 | 265 | 9055797 | ||
BC145504.1 | - | 2, 3 | 265 | 9054067 |
