siRNA Details
Gene Symbol: Lrig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177152.5 | NP_796126.4 | 19 | 3537 | ||
GenBank mRNA | AK018522.1 | - | 13 | 2350 | ||
AK020260.1 | - | 2 | 3118 | |||
AK078783.1 | - | 5 | 1131 | |||
AK129481.1 | - | 15 | 2486 | |||
AK135388.1 | - | 3 | 1169 | |||
AY505341.1 | - | 19 | 3480 | |||
BC038142.1 | - | 2 | 213 | |||
BC065142.1 | - | 19 | 3503 | 30610561 |
