siRNA Details
Gene Symbol: Ubn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177185.4 | NP_796159.3 | 14 | 3156 | ||
GenBank mRNA | AK044162.1 | - | 263 | |||
AK051609.1 | - | 14 | 2529 | |||
AK173339.1 | - | 2 | 4192 | |||
BC043095.1 | - | 2452 | ||||
BC051458.1 | - | 14 | 3128 | 6468017 |
