siRNA Details
Gene Symbol: Ccdc169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290138.1 | NP_001277067.1 | 7 | 1191 | ||
NM_001290141.1 | NP_001277070.1 | 7 | 666 | |||
NM_177203.3 | NP_796177.2 | 7 | 570 | |||
GenBank mRNA | AK042903.1 | - | 7 | 1193 | ||
BC150827.1 | - | 6 | 388 | |||
BC171976.1 | - | 7 | 425 | |||
BC171978.1 | - | 8 | 459 | |||
BC171981.1 | - | 6 | 388 | |||
BU962217.1 | - | 572 |
