siRNA Details
Gene Symbol: D5Ertd579e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081232.3 | NP_001074701.3 | 6 | 2757 | ||
GenBank mRNA | AK122224.1 | - | 5 | 2268 | ||
AK134363.1 | - | 6 | 2718 | |||
AK139370.1 | - | 6 | 2674 | |||
BC094673.1 | - | 1 | 1290 | |||
BC151085.1 | - | 5 | 2502 | 9087998 |
