siRNA Details
Gene Symbol: D5Ertd579e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081232.3 | NP_001074701.3 | 6 | 3004 | ||
GenBank mRNA | AK122224.1 | - | 5 | 2515 | ||
AK134363.1 | - | 6 | 2965 | |||
AK139370.1 | - | 6 | 2921 | |||
BC094673.1 | - | 1 | 1537 | |||
BC151085.1 | - | 5 | 2749 | 9087998 |
