siRNA Details
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146043.2 | NP_001139515.1 | 19 | 2057 | ||
NM_001271463.1 | NP_001258392.1 | 19 | 2057 | |||
NM_001271464.1 | NP_001258393.1 | 18 | 1976 | |||
NM_001271465.1 | NP_001258394.1 | 19 | 2084 | |||
NM_177266.5 | NP_796240.3 | 19 | 2090 | |||
GenBank mRNA | AK035819.1 | - | 19 | 1995 | ||
AK044371.1 | - | 18 | 1977 | |||
AK052633.1 | - | 3 | 400 | |||
AK083441.1 | - | 18 | 2096 | |||
AK161797.1 | - | 19 | 1993 | |||
AK165871.1 | - | 19 | 2051 | |||
BC027341.1 | - | 19 | 2041 | 3984129 | ||
BC057062.1 | - | 8 | 1868 | |||
BC138233.1 | - | 19 | 1988 | 8861252 | ||
BC145208.1 | - | 19 | 1988 | 9053759 |
