siRNA Details
Gene Symbol: Ep300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177821.6 | NP_808489.4 | 22, 23 | 4227 | ||
GenBank mRNA | AK040384.1 | - | 2, 3 | 175 | ||
BC144976.1 | - | 22, 23 | 4081 | 9053524 | ||
BC150681.1 | - | 22, 23 | 4081 | 9087592 | ||
CD348567.1 | - | 1 | 20 | |||
CN535246.1 | - | 7, 8 | 745 | |||
CN536119.1 | - | 5, 6 | 384 |
