siRNA Details
Gene Symbol: Ep300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177821.6 | NP_808489.4 | 16, 17 | 3543 | ||
GenBank mRNA | AK139715.1 | - | 13, 14 | 2036 | ||
BC144976.1 | - | 16, 17 | 3397 | 9053524 | ||
BC150681.1 | - | 16, 17 | 3397 | 9087592 | ||
BY759831.1 | - | 1, 2 | 134 | |||
CN535246.1 | - | 1, 2 | 61 | |||
CN537190.1 | - | 2, 3 | 248 |
