siRNA Details
Gene Symbol: Dennd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166501.1 | NP_001159973.1 | 5 | 366 | ||
NM_181347.3 | NP_851992.1 | 4 | 232 | |||
GenBank mRNA | AK155727.1 | - | 4 | 233 | ||
BC042698.1 | - | 5 | 289 | |||
BC141388.1 | - | 4 | 228 | 9056001 | ||
BC141389.1 | - | 4 | 228 | 9056002 |
