siRNA Details
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033285.1 | NP_001028457.1 | 4 | 458 | ||
GenBank mRNA | AK138649.1 | - | 5 | 947 | ||
AK162174.1 | - | 4 | 356 | |||
BC158017.1 | - | 4 | 458 | 9007323 | ||
BC158095.1 | - | 4 | 458 | 9007382 |
