siRNA Details
Gene Symbol: Acot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025590.4 | NP_079866.2 | 13 | 1399 | ||
GenBank mRNA | AF416923.1 | - | 13 | 1337 | ||
AK031588.1 | - | 13 | 1316 | |||
AK038546.1 | - | 13 | 1337 | |||
AK048670.1 | - | 13 | 1399 | |||
AK052332.1 | - | 13 | 1415 | |||
AK122350.1 | - | 12 | 1186 | |||
AK170189.1 | - | 6 | 422 | |||
AK171255.1 | - | 6 | 423 | |||
BC042492.1 | - | 13 | 1334 | 4240966 |
