siRNA Details
Gene Symbol: Acot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025590.4 | NP_079866.2 | 1 | 132 | ||
GenBank mRNA | AF416923.1 | - | 1 | 70 | ||
AK031588.1 | - | 1 | 49 | |||
AK048670.1 | - | 1 | 132 | |||
AK052332.1 | - | 1 | 148 | |||
BC042492.1 | - | 1 | 67 | 4240966 |
