siRNA Details
Gene Symbol: Kctd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159941.1 | NP_001153413.1 | 5, 6 | 608 | ||
NM_026145.4 | NP_080421.2 | 5, 6 | 605 | |||
GenBank mRNA | AK028657.1 | - | 5, 6 | 572 | ||
AK041516.1 | - | 5, 6 | 572 | |||
AK145295.1 | - | 5, 6 | 584 | |||
AK147714.1 | - | 5, 6 | 550 | |||
AK161199.1 | - | 5, 6 | 562 | |||
AK163351.1 | - | 5, 6 | 559 | |||
AK166505.1 | - | 5, 6 | 584 | |||
AK216751.1 | - | 2, 3 | 98 | |||
BC006935.1 | - | 5, 6 | 586 | 3601917 | ||
BQ571808.1 | - | 5, 6 | 559 |
