siRNA Details
Gene Symbol: Kctd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159941.1 | NP_001153413.1 | 4 | 537 | ||
NM_026145.4 | NP_080421.2 | 4 | 534 | |||
GenBank mRNA | AK028657.1 | - | 4 | 501 | ||
AK041516.1 | - | 4 | 501 | |||
AK145295.1 | - | 4 | 513 | |||
AK147714.1 | - | 4 | 479 | |||
AK161199.1 | - | 4 | 491 | |||
AK163351.1 | - | 4 | 488 | |||
AK166505.1 | - | 4 | 513 | |||
AK216751.1 | - | 1 | 27 | |||
BC006935.1 | - | 4 | 515 | 3601917 | ||
BQ571808.1 | - | 4 | 488 |
