siRNA Details
Gene Symbol: Arhgap32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195632.1 | NP_001182561.1 | 18 | 2025 | ||
NM_177379.4 | NP_796353.3 | 9 | 1361 | |||
GenBank mRNA | AB088417.1 | - | 10 | 1308 | ||
AY194286.1 | - | 9 | 1088 | |||
BC132390.1 | - | 8 | 1017 | 40130667 | ||
BC138042.1 | - | 8 | 1017 | 8861061 |
