siRNA Details
Gene Symbol: Hcn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081192.1 | NP_001074661.1 | 2 | 1031 | ||
GenBank mRNA | AF064874.1 | - | 2 | 498 | ||
BC158106.1 | - | 2 | 1131 | 9088182 | ||
BC158108.1 | - | 2 | 1131 | 9088183 |
