siRNA Details
Gene Symbol: Tgm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177911.4 | NP_808579.2 | 10 | 1211 | ||
GenBank mRNA | AF486627.1 | - | 10 | 1210 | ||
AK035279.1 | - | 10 | 1212 | |||
BC141297.1 | - | 10 | 1208 | 9055909 | ||
BC145622.1 | - | 10 | 1208 | 9054190 |
