siRNA Details
Gene Symbol: Tgm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177911.4 | NP_808579.2 | 4 | 398 | ||
GenBank mRNA | AF486627.1 | - | 4 | 397 | ||
AK035279.1 | - | 4 | 399 | |||
BC141297.1 | - | 4 | 395 | 9055909 | ||
BC145622.1 | - | 4 | 395 | 9054190 |
