siRNA Details
Gene Symbol: Gm5127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033541.2 | NP_001028713.1 | 4 | 943 | ||
GenBank mRNA | AK044840.1 | - | 7 | 1285 | ||
AK142438.1 | - | 4 | 944 | |||
BC147417.1 | - | 1 | 618 | 9056484 | ||
BC147418.1 | - | 1 | 618 | 9056504 |
