siRNA Details
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177381.3 | NP_796355.2 | 17 | 1919 | ||
GenBank mRNA | AK140362.1 | - | 17 | 1919 | ||
BC038030.1 | - | 18 | 1930 | 4020725 | ||
BC048005.1 | - | 13 | 1274 | |||
CO432313.1 | - | 4 | 355 |

More Information
Other Species Targeted

Gene Ontology Categories:
