siRNA Details
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177381.3 | NP_796355.2 | 13 | 1511 | ||
GenBank mRNA | AK140362.1 | - | 13 | 1511 | ||
BC038030.1 | - | 14 | 1522 | 4020725 | ||
BC048005.1 | - | 9 | 866 |
