siRNA Details
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177381.3 | NP_796355.2 | 10 | 1052 | ||
GenBank mRNA | AK140362.1 | - | 10 | 1052 | ||
BC038030.1 | - | 11 | 1063 | 4020725 | ||
BC048005.1 | - | 6 | 407 |
