siRNA Details
Gene Symbol: Morc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045529.3 | NP_001038994.2 | 16 | 2728 | ||
GenBank mRNA | AK168211.1 | - | 7 | 1414 | ||
AK172896.1 | - | 16 | 2728 | |||
BC026506.1 | - | 3 | 1081 | |||
BC029284.1 | - | 3 | 958 | |||
BC098072.1 | - | 16 | 2569 | 30109159 | ||
BC145705.1 | - | 16 | 2677 | 40131008 |
