siRNA Details
Gene Symbol: Morc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045529.3 | NP_001038994.2 | 13, 14 | 1656 | ||
GenBank mRNA | AK132226.1 | - | 12, 13 | 1616 | ||
AK168211.1 | - | 4, 5 | 343 | |||
AK172896.1 | - | 13, 14 | 1656 | |||
BC098072.1 | - | 13, 14 | 1497 | 30109159 | ||
BC145705.1 | - | 13, 14 | 1605 | 40131008 |
