siRNA Details
Gene Symbol: Myo18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291212.1 | NP_001278141.1 | 40 | 6059 | ||
NM_001291213.1 | NP_001278142.1 | 40 | 6167 | |||
NM_001291214.1 | NP_001278143.1 | 38 | 5014 | |||
NM_001291215.1 | NP_001278144.1 | 41 | 5080 | |||
NM_011586.3 | NP_035716.1 | 39 | 6023 | |||
GenBank mRNA | AB026497.1 | - | 39 | 5993 | ||
AK147584.1 | - | 41 | 5029 | |||
AK171342.1 | - | 38 | 5011 | |||
AY692137.1 | - | 3 | 191 | |||
AY692138.1 | - | 3 | 191 | |||
AY692139.1 | - | 2 | 146 | |||
BC046638.1 | - | 22 | 2851 | |||
BC057920.1 | - | 10 | 1234 | |||
BC138366.1 | - | 40 | 6001 | 8861386 | ||
BC145257.1 | - | 40 | 6109 |
