siRNA Details
Gene Symbol: Tmem237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033449.1 | NP_001028621.1 | 9 | 902 | ||
NM_001037812.2 | NP_001032901.1 | 9 | 861 | |||
GenBank mRNA | AK133112.1 | - | 10 | 886 | ||
AK161697.1 | - | 9 | 905 | |||
AK167733.1 | - | 9 | 863 | |||
BC147243.1 | - | 9 | 881 | 9056304 | ||
BC147244.1 | - | 9 | 881 | 9056305 | ||
CA465507.1 | - | 159 |
