siRNA Details
Gene Symbol: Gm996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005424.2 | NP_001005424.2 | 1 | 2235 | ||
GenBank mRNA | BC065698.1 | - | 2 | 2136 | 6825178 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006233653, XM_006233652, XM_008761615, FQ211897.
Gene Ontology Categories:
