siRNA Details
Gene Symbol: Tnrc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122730.2 | NP_001116202.2 | 23 | 6865 | ||
GenBank mRNA | BC094365.1 | - | 4 | 394 | ||
CB244319.1 | - | 6 | 620 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tnrc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122730.2 | NP_001116202.2 | 23 | 6865 | ||
GenBank mRNA | BC094365.1 | - | 4 | 394 | ||
CB244319.1 | - | 6 | 620 |