siRNA Details
Gene Symbol: Ccdc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029212.3 | NP_083488.1 | 5, 6 | 815 | ||
GenBank mRNA | AB048860.1 | - | 5, 6 | 889 | ||
BC116291.1 | - | 5, 6 | 815 | 40094912 | ||
BC116292.1 | - | 5, 6 | 815 | 40094918 |
