siRNA Details
Gene Symbol: Cep162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199316.2 | NP_955020.2 | 23 | 3791 | ||
GenBank mRNA | BC150998.1 | - | 23 | 3596 | 9087911 | |
BC151006.1 | - | 23 | 3596 | 9087919 | ||
BC151104.1 | - | 23 | 3596 | 9088017 | ||
CB246096.1 | - | 1 | 362 |
