siRNA Details
Gene Symbol: Zfp667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024928.2 | NP_001020099.1 | 5 | 1428 | ||
GenBank mRNA | AK164056.1 | - | 6 | 1337 | ||
AY850275.1 | - | 5 | 1428 | |||
BC141152.1 | - | 5 | 1463 | 9055764 | ||
BC141153.1 | - | 5 | 1463 | 9055765 |
