siRNA Details
Gene Symbol: BC067074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC067074.1 | - | 1, 2 | 31 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC067074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC067074.1 | - | 1, 2 | 31 |