siRNA Details
Gene Symbol: Itprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001738.2 | NP_001001738.2 | 2 | 1354 | ||
GenBank mRNA | AK160686.1 | - | 2 | 1356 | ||
AK165192.1 | - | 2 | 1333 | |||
AK171522.1 | - | 2 | 1260 | |||
AK171539.1 | - | 2 | 1260 | |||
AK171911.1 | - | 2 | 1335 | |||
AK220222.1 | - | 2 | 1268 | |||
BC063749.1 | - | 3 | 1320 | 6813779 |
