siRNA Details
Gene Symbol: Gnptab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004164.2 | NP_001004164.2 | 10 | 1201 | ||
GenBank mRNA | AK140867.1 | - | 9 | 1305 | ||
AK154710.1 | - | 2 | 100 | |||
AK173132.1 | - | 10 | 1201 | |||
BC060638.1 | - | 11 | 1309 | 6849405 |
