siRNA Details
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033488.2 | NP_001028660.2 | 4 | 427 | ||
GenBank mRNA | AK133061.1 | - | 4 | 442 | ||
AK156138.1 | - | 5 | 579 | |||
AK157892.1 | - | 3 | 1254 | |||
AK162620.1 | - | 4 | 426 | |||
AK172423.1 | - | 4 | 428 | |||
BC145584.1 | - | 4 | 440 |
