siRNA Details
Gene Symbol: Gm5512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CK334097.1 | - | 1 | 59 |
Gene Symbol: Rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025343.5 | NP_079619.4 | 3 | 286 | ||
GenBank mRNA | AK007323.1 | - | 3 | 234 | ||
AK075603.1 | - | 3 | 269 | |||
AK169025.1 | - | 3 | 279 | |||
AK180283.1 | - | 243 | ||||
AK182595.1 | - | 2 | 182 | |||
BC027299.1 | - | 3 | 234 | 3987394 | ||
BC036150.1 | - | 2 | 206 | 4977394 | ||
BY740212.1 | - | 3 | 279 | |||
CJ184230.1 | - | 3 | 287 |
