siRNA Details
Gene Symbol: Rnf207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033489.2 | NP_001028661.2 | 14 | 1682 | ||
GenBank mRNA | AK042379.1 | - | 14 | 1665 | ||
BC141256.1 | - | 14 | 1566 | 9055868 | ||
BC141257.1 | - | 14 | 1566 | 9055869 |
