siRNA Details
Gene Symbol: Fam92b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033980.2 | NP_001029152.2 | 3, 4 | 467 | ||
GenBank mRNA | AK131790.1 | - | 3, 4 | 467 | ||
BC147233.1 | - | 3, 4 | 386 | 9056283 | ||
BC147234.1 | - | 3, 4 | 386 | 9056269 | ||
BC172014.1 | - | 2, 3 | 151 |
