siRNA Details
Gene Symbol: Nox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285833.1 | NP_001272762.1 | 10, 11 | 1206 | ||
NM_001285835.1 | NP_001272764.1 | 10, 11 | 1206 | |||
NM_015760.5 | NP_056575.1 | 10, 11 | 1210 | |||
GenBank mRNA | AB041034.1 | - | 10, 11 | 1061 | ||
AB042745.1 | - | 10, 11 | 1061 | |||
AF261944.1 | - | 10, 11 | 1065 | |||
AK050371.1 | - | 10, 11 | 1065 | |||
AK052905.1 | - | 10, 11 | 1061 | |||
AK085509.1 | - | 10, 11 | 1066 | |||
AK149398.1 | - | 10, 11 | 1061 | |||
AK149455.1 | - | 10, 11 | 1061 | |||
AK149492.1 | - | 10, 11 | 1061 |
