siRNA Details
Gene Symbol: Txnrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042513.1 | NP_001035978.1 | 9 | 1077 | ||
NM_001042514.1 | NP_001035979.1 | 8 | 1165 | |||
NM_001042523.1 | NP_001035988.1 | 9 | 1294 | |||
NM_015762.2 | NP_056577.2 | 8 | 970 | |||
GenBank mRNA | AB027565.1 | - | 8 | 997 | ||
AF333036.1 | - | 9 | 1294 | |||
AK011902.1 | - | 9 | 1078 | |||
AK146125.1 | - | 8 | 971 | |||
AK149625.1 | - | 9 | 1078 | |||
AK151927.1 | - | 9 | 1078 | |||
AK168356.1 | - | 9 | 1078 | |||
AK172383.1 | - | 7 | 3330 | |||
AK181192.1 | - | 2 | 179 | |||
AK183038.1 | - | 2 | 179 | |||
AK185436.1 | - | 2 | 179 | |||
AK189898.1 | - | 2 | 179 | |||
AK192872.1 | - | 2 | 179 | |||
AK195172.1 | - | 2 | 178 | |||
AK198860.1 | - | 2 | 179 | |||
AK203979.1 | - | 2 | 179 | |||
AK204938.1 | - | 2 | 179 | |||
BC020274.1 | - | 8 | 932 | |||
BC037643.1 | - | 8 | 1250 | 5059831 |
