siRNA Details
Gene Symbol: Txnrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042513.1 | NP_001035978.1 | 8 | 879 | ||
NM_001042514.1 | NP_001035979.1 | 7 | 967 | |||
NM_001042523.1 | NP_001035988.1 | 8 | 1096 | |||
NM_015762.2 | NP_056577.2 | 7 | 772 | |||
GenBank mRNA | AB027565.1 | - | 7 | 799 | ||
AF333036.1 | - | 8 | 1096 | |||
AK011902.1 | - | 8 | 880 | |||
AK146125.1 | - | 7 | 773 | |||
AK149625.1 | - | 8 | 880 | |||
AK151927.1 | - | 8 | 880 | |||
AK168356.1 | - | 8 | 880 | |||
AK172383.1 | - | 6 | 1205 | |||
BC020274.1 | - | 7 | 734 | |||
BC037643.1 | - | 7 | 1052 | 5059831 |
