siRNA Details
Gene Symbol: Txnrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042513.1 | NP_001035978.1 | 6 | 643 | ||
NM_001042514.1 | NP_001035979.1 | 5 | 731 | |||
NM_001042523.1 | NP_001035988.1 | 6 | 860 | |||
NM_015762.2 | NP_056577.2 | 5 | 536 | |||
GenBank mRNA | AB027565.1 | - | 5 | 563 | ||
AF333036.1 | - | 6 | 860 | |||
AK011902.1 | - | 6 | 644 | |||
AK146125.1 | - | 5 | 537 | |||
AK149625.1 | - | 6 | 644 | |||
AK151927.1 | - | 6 | 644 | |||
AK168356.1 | - | 6 | 644 | |||
AK172383.1 | - | 5 | 569 | |||
AK208113.1 | - | 1 | 45 | |||
AK210721.1 | - | 2 | 233 | |||
AK210722.1 | - | 2 | 234 | |||
AK212800.1 | - | 2 | 234 | |||
AK217451.1 | - | 1 | 45 | |||
AK218801.1 | - | 1 | 45 | |||
BC020274.1 | - | 5 | 498 | |||
BC037643.1 | - | 5 | 816 | 5059831 | ||
CX562215.1 | - | 5 | 731 |
